Learn More
TBCD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560946
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: FQETDKAWHG GCLALAELGR RGLLLPSRLV DVVAVILKAL TYDEKRGACS VGTNVRDAAC YVCWAFARAY EPQELKPFVT AISSALVIAA VFDRDINCRR AASAAFQENV GRQGTFPHGI DILTTADYFA VGN Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Cofactor D is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.Spezifikation
TBCD | |
Polyclonal | |
Unconjugated | |
Tbcd | |
2310057L06Rik; A030005L14Rik; Beta-tubulin cofactor D; KIAA0988; mKIAA0988; PP1096; SSD1; SSD-1; TBCD; TFCD; tubulin folding cofactor D; tubulin-folding cofactor D; Tubulin-specific chaperone D | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6904 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BTW9 | |
Tbcd | |
Recombinant protein corresponding to Human TBCD | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only