Learn More
Tocris Bioscience™ TC NTR1 17
Selective non-peptide NTS1 agonist
N.V.
Spezifikation
Produkttyp | TC NTR1 17 |
---|---|
Beschreibung | TC NTR1 17 |
Molekulargewicht | 522.98 |
Beschreibung
Non-peptide neurotensin receptor 1 (NTS1) partial agonist (EC50 <15.6 nM and Emax = 63% of neurotensin response in Ca2+ mobilization assay). Exhibits over 50-fold selectivity for NTS1 over NTS2 and GPR35.Spezifikation
TC NTR1 17 | |
TC NTR1 17 | |
522.98 |
For Research Use Only