Learn More
TCEAL8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563658
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: PQNMPKAEED RPLEDVPQEA EGNPQPSEEG VSQEAEGNPR GGPNQPGQGF KEDTPVRHLD PEEMIRGVDE LERLREEIRR VRNKFVMMHW KQRHSRSRPY PVCFR Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 71%.
TCEAL8 Polyclonal antibody specifically detects TCEAL8 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
TCEAL8 | |
Polyclonal | |
Unconjugated | |
TCEAL8 | |
3930402F23Rik; MGC45400; TCEAL8; TCEA-like protein 8; transcription elongation factor A (SII)-like 8; transcription elongation factor A like 8; transcription elongation factor A protein-like 8; Transcription elongation factor S-II protein-like 8; up-regulated in nephrectomized rat kidney #1; UR-NR#1; WEX3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90843 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IYN2 | |
TCEAL8 | |
Recombinant protein corresponding to Human TCEAL8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only