Learn More
TCHHL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560032
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KHSNIQEPPL QREDEPSSQH ADLPEQAAAR SPSQTQKSTD SKDVCRMFDT QEPGKDADQT PAKTKNLGEP EDYGRTSETQ EKECETKDLP VQYGSRN Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 38%.
This gene belongs to the S100 fused-type protein (SFTP) gene family, and is located in a cluster of SFTP genes on chromosome 1q21. Several members of this family have been implicated in the development of complex skin disorders. This gene is evolutionarily conserved; its expression appears to be hair-specific and spatially restricted within the distal inner root sheath of the hair follicle. It thus may have an important role in hair morphogenesis. [provided by RefSeq, Aug 2013].Spezifikation
TCHHL1 | |
Polyclonal | |
Unconjugated | |
TCHHL1 | |
basalin; intermediate filament-associated protein; Protein S100-A17; S100 calcium binding protein A17; S100 calcium-binding protein A17; S100A17; TCHHL1; THHL1; trichohyalin like 1; trichohyalin-like 1; trichohyalin-like protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126637 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5QJ38 | |
TCHHL1 | |
Recombinant protein corresponding to Human TCHHL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only