missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561708
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LILIEKELAE LGWKFLNLMH HNQQVFGPYY AEILKH Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 81%.
Plays a role in the process of sperm capacitation and acrosome reactions. Probable receptor for the putative fertilization-promoting peptide (FPP) at the sperm membrane that may modulate the activity of the adenylyl cyclase cAMP pathway. [UniProt]Spezifikation
TCP11 | |
Polyclonal | |
Unconjugated | |
TCP11 | |
D17Ken1; D6S230E; fertilization-promoting peptide receptor; FPPR; t-complex 11; t-complex 11 (a murine tcp homolog); t-complex 11 homolog; t-complex 11, testis-specific; t-complex protein 11; T-complex protein 11 homolog; Tcp11; Tcp-11; testis secretory sperm-binding protein Li 222n; testis-specific protein PBS13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6954 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WWU5 | |
TCP11 | |
Recombinant protein corresponding to Human TCP11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only