Learn More
TCP11L2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559905
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SLIDKRIKLY MRRLLCLPSP QKCMPPMPGG LAVIQQELEA LGSQYANIVN LNKQVYGPFY ANILRKLLFN EEAMGKVDAS PPTN Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 80%.
TCP11L2 Polyclonal antibody specifically detects TCP11L2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TCP11L2 | |
Polyclonal | |
Unconjugated | |
TCP11L2 | |
t-complex 11 like 2; t-complex 11, testis-specific-like 2; T-complex protein 11-like protein 2; TCP11L2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
255394 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N4U5 | |
TCP11L2 | |
Recombinant protein corresponding to Human TCP11L2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only