missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TCR V beta 11 Monoclonal Antibody (RR3-15), APC, eBioscience™
GREENER_CHOICE

Artikelnummer. p-7178801
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
15558136 100 μg 100 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15558136 Lieferant Invitrogen™ Lieferanten-Nr. 17582782

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rat Monoclonal Antibody

Description: This RR3-15 monoclonal antibody reacts with the mouse T-cell receptor (TCR) V beta 11 chain. Composed of an alpha and beta chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The RR3-15 antibody recognizes the V beta 11 chain on T cells from mouse strains with the b haplotype in the Tcrb gene complex, including C57BL/6 and B10, but is absent in mice with the a haplotype, such as C57L, SJL, and SWR. In addition, reports indicate that Vbeta11+ T cells are eliminated in mice bearing the MHC Class II I-E molecule. Finally, V beta 11+CD4+ T cells have been associated with graft-versus-host disease (GVHD) severity. The RR3-15 antibody has been reported to activate V beta 11+ T cells. Applications Reported: This RR3-15 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RR3-15 antibody has been tested by flow cytometric analysis of mouse lymph node cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease. Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen TCR V beta 11
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Klon RR3-15
Konzentration 0.2 mg/mL
Konjugat APC
Zusammensetzung PBS with 0.09% sodium azide; pH 7.2
Gen-Alias TCR V beta11; TCRV beta 11; TCRV beta11; Vb11; Vbeta11
Wirtsspezies Rat
Reinigungsverfahren Affinity chromatography
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Zielspezies Mouse
Inhalt und Lagerung 4°C, store in dark, DO NOT FREEZE!
Produkttyp Antibody
Form Liquid
Isotype IgG2b κ
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.