missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR V beta 11 Monoclonal Antibody (RR3-15), Super Bright™ 436, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Marke: Affymetrix eBioscience 62-5827-82
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Rat Monoclonal Antibody
TCR V beta 11 Monoclonal antibody specifically detects TCR V beta 11 in Mouse samples. It is validated for Flow Cytometry.Spezifikation
TCR V beta 11 | |
Monoclonal | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Rat | |
100 μg | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
RR3-15 | |
Super Bright 436 | |
TCR V beta11; TCRV beta 11; TCRV beta11; Vb11; Vbeta11 | |
Affinity chromatography | |
RUO | |
Mouse | |
Antibody | |
IgG2b κ |