Learn More
TCR V delta 1 Monoclonal Antibody (TS8.2), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Marke: Affymetrix eBioscience 17-5679-42
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Description: This TS8.2 monoclonal antibody recognizes human TCR V delta 1, a common framework determinant on the delta-subunit of the T cell receptor. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity. TCR V delta 1 Monoclonal Antibody (TS8.2) has been validated for the following applications: Flow Cytometry Applications Reported: This TS8.2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TS8.2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser
TCR V delta 1 Monoclonal antibody specifically detects TCR V delta 1 in Human samples. It is validated for Flow Cytometry.Spezifikation
TCR V delta 1 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
0 | |
Mouse | |
Affinity chromatography | |
RUO | |
6964 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
TS8.2 | |
APC | |
TRD | |
TRD | |
Full length protein of Human TCR V delta 1 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |