Learn More
TDRD5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA556139
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: AMANHDIPPD AVPNKKLCRL PPLDTSSLIG VFVEYIISPS QFYIRIYSRD SSELLEDMMI EMRRCYSNQL VSDRYVMPEC FIQPGHLCCV RISEDKWWYR VIIHRVLEKQ EVEVFYPDFG NIGIVQKSSL RFLKCCYTK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
TDRD5 Polyclonal antibody specifically detects TDRD5 in Human samples. It is validated for Immunohistochemistry.Spezifikation
TDRD5 | |
Polyclonal | |
Unconjugated | |
TDRD5 | |
Gm103; Tdrd5; testicular tissue protein Li 194; tudor domain containing 5; tudor domain containing protein 5; tudor domain-containing protein 5; TUDOR3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
163589 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NAT2 | |
TDRD5 | |
Recombinant protein corresponding to Human TDRD5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only