missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TEC, His Tag Recombinant Protein
Marke: Invitrogen PV3269
Weitere Details : Netto-Gewicht : 3.36000kg
Beschreibung
TEC is a non-receptor protein-tyrosine kinase containing a pleckstrin homology domain and is highly expressed in many hematopoietic cell lines.Spezifikation
NP_003206 | |
7006 | |
10 μg | |
TEC Kinase Family | |
Recombinant | |
Storage Buffer: 20 mM Tris, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.05% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate | |
tec protein tyrosine kinase | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
76.4 kDa | |
TEC, PSCTK4 | |
Human | |
His Tag | |
Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
TEC | |
Baculovirus | |
Full Length, Recombinant, Cytoplasmic | |
Cytoplasmic Tyrosine Kinases |