Learn More
TER-119 Monoclonal Antibody (TER-119), Super Bright™ 702, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
166.00 € - 336.00 €
Spezifikation
Antigen | TER-119 |
---|---|
Klon | TER-119 |
Konzentration | 0.2 mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15903774
|
Affymetrix eBioscience
67-5921-80 |
25 μg |
166.00 €
25 Mikrogramm |
Verfügbar ab: 14-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15913774
|
Affymetrix eBioscience
67-5921-82 |
100 μg |
336.00 €
100 Mikrogramm |
Verfügbar ab: 14-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Description: The RB6-8C5 monoclonal antibody reacts with mouse Ly-6G, a 21-25 kDa protein also known as the myeloid differentiation antigen Gr-1. A GPI-linked protein, Gr-1 is expressed by the myeloid lineage in a developmentally regulated manner in the bone marrow. While monocytes only express Gr-1 transiently during their bone marrow development, the expression of Gr-1 on bone marrow granulocytes as well as on peripheral neutrophils is a good marker for these populations. eBioscience testing indicates that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also stains cells that express the highest levels of Ly6c (as defined by staining with antibody clone HK1.4). It is recommended that 1A8-Ly6G (cat. 9668) be used when looking at Ly-6G specific targets. Applications Reported: This RB6-8C5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RB6-8C5 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 702 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 702 nm.
TER-119 Monoclonal antibody specifically detects TER-119 in Mouse samples. It is validated for Flow Cytometry.Spezifikation
TER-119 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Ly76; Ly-76; TER119 | |
Ly76 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Ly76 |
TER-119 | |
Flow Cytometry | |
Super Bright 702 | |
Rat | |
Mouse | |
0 | |
104231 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |