missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-Aminobenzoate, TRC

Artikelnummer. 30415198
Change view
Click to view available options
Menge:
5 mg
10 mg
50 mg
Unit Size:
5 Milligramm
50 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30415198 10 mg -
30411107 50 mg 50 Milligramm
30409761 5 mg 5 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30415198 Supplier Toronto Research Chemicals Supplier No. TRCT20584310MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64113-91-3
Summenformel C11H15NO2
Molekulargewicht (g/mol) 193.24
IUPAC-Name tert-butyl 2-aminobenzoate
SMILES CC(C)(C)OC(=O)c1ccccc1N

Specifications

Chemischer Name oder Material tert-Butyl 2-Aminobenzoate
Menge 10 mg
InChI-Formel InChI=1S/C11H15NO2/c1-11(2,3)14-10(13)8-6-4-5-7-9(8)12/h4-7H,12H2,1-3H3
Formelmasse 193.1103
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.