missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosteronbenzoat 1,0 mg/ml in Acetonitril, LoGiCal
Spezifikation
Specifications
| Chemischer Name oder Material | Testosterone Benzoate |
| CAS | 2088-71-3 |
| Bezeichnung der Analyten oder Bestandteile | Testosterone Benzoate |
| Versandbedingung | Room Temperature |
| Summenformel | C26 H32 O3 |
| Zertifizierung/Konformität | ISO 17034 |
| Synonym | Testosterone benzoate, Testosterone, benzoate (7CI,8CI), NSC 524278, Testosterone 17-benzoate, Androst-4-en-3-one, 17-(benzoyloxy)-, (17beta)-, (17beta)-17-(Benzoyloxy)androst-4-en-3-one, 17beta-Benzoyloxyandrost-4-en-3-one, 17beta-Hydroxyandrost-4-en-3-one benzoate, Androst-4-en-17beta-ol-3-one benzoate |
| InChI-Formel | InChI=1S/C26H32O3/c1-25-14-12-19(27)16-18(25)8-9-20-21-10-11-23(26(21,2)15-13-22(20)25)29-24(28)17-6-4-3-5-7-17/h3-7,16,20-23H,8-15H2,1-2H3/t20-,21-,22-,23-,25-,26-/m0/s1 |
| Lösungstyp | Single Solution |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2OC(=O)c5ccccc5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?