Learn More
Tetrahydropyran-2-carbonsäure, 97 %, Thermo Scientific™
Catalog Number p-3830472
Click to view available options
Menge:
250 mg
1 g
51673-83-7
C6H10O3
130.143
MFCD07779239
MQAYFGXOFCEZRW-UHFFFAOYSA-N
tetrahydro-2h-pyran-2-carboxylic acid, tetrahydropyran-2-carboxylic acid, 2h-pyran-2-carboxylic acid, tetrahydro, 2h-3,4,5,6-tetrahydropyran-2-carboxylic acid, pubchem19589, acmc-20m8cx, acmc-20m8cy, 2-tetrahydropyranoic acid, tetrahydro-pyran-carboxylic acid, tetrahydro-pyran-2-carboxylic acid
10964532
Oxan-2-Carbonsäure
C1CCOC(C1)C(=O)O
51673-83-7
C6H10O3
130.143
MFCD07779239
MQAYFGXOFCEZRW-UHFFFAOYSA-N
tetrahydro-2h-pyran-2-carboxylic acid, tetrahydropyran-2-carboxylic acid, 2h-pyran-2-carboxylic acid, tetrahydro, 2h-3,4,5,6-tetrahydropyran-2-carboxylic acid, pubchem19589, acmc-20m8cx, acmc-20m8cy, 2-tetrahydropyranoic acid, tetrahydro-pyran-carboxylic acid, tetrahydro-pyran-2-carboxylic acid
10964532
Oxan-2-Carbonsäure
C1CCOC(C1)C(=O)O
Chemischer Name oder Material Tetrahydropyran-2-carboxylic acid
CAS Min. % 96.5
CAS Max. % 100.0
Prozentgehaltsbereich 95%
Menge 250 mg
Formelmasse 130.14
Reinheit (%) 95%