Learn More
Tetramethylsilane 25 mL, Eurisotop
Specifications
Specifications
| CAS | 75-76-3 |
| Color | Colorless |
| Quantity | 25 mL |
| Molecular Formula | C4H12Si |
| MDL Number | MFCD00008274 |
| Synonym | Tetramethylsilane |
| InChI Key | CZDYPVPMEAXLPK-UHFFFAOYSA-N |
| SMILES | C[Si](C)(C)C |
| IUPAC Name | tetramethylsilane |
| Molecular Weight (g/mol) | 88.23 |
| Show More |