Learn More
Sino Biological™ TF (Transferrin) Recombinant Human Protein, His Tag
Expressed from DNA sequence encoding human TF (Met 1-Pro 698) fused to polyhistidine tag at C-terminus
Marke: Sino Biological™ 11019H08H25
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Expressed from DNA sequence encoding human TF (Met 1-Pro 698) fused to polyhistidine tag at C-terminus
- Measured by its binding ability in functional ELISA
- Immobilized human CD71 at 10 μg/mL(100μgL/well) binds human Transferrin
- EC50 of human Transferrin is 5.6ng/mL
- Lyophilized in 25mM Tris, 150mM NaCl, pH 7.5, 5% Mannit
Spezifikation
Protein Structure Analysis, Assay Standard | |
5 x 5μg | |
Full Length, Recombinant | |
Transferrin | |
TF (Transferrin) Recombinant Human Protein | |
Mammalian | |
Lyophilized |
76.6kd | |
Human | |
<1 EU/μg | |
TF | |
His Tag | |
Other proteins | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.