missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGM4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15847-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
TGM4 is associated with the mammalian reproductive process. TGM4 catalyzes the cross-linking of proteins and the conjugation of polyamines to specific proteins in the seminal tract.Spezifikation
TGM4 | |
Polyclonal | |
Unconjugated | |
TGM4 | |
Fibrinoligase, hTGP, Prostate transglutaminase, TG(P), TGase 4, TGase P, TGM4, TGP, Transglutaminase 4, transglutaminase 4 (prostate), Transglutaminase P | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
331046, 7047 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P49221, Q8BZH1 | |
Tgm4 | |
TGM4 Fusion Protein Ag8608 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |