Learn More
Invitrogen™ Human GLRX2, Recombinant Protein
Recombinant Protein
Marke:  Invitrogen™ LFP0016
Weitere Details : Netto-Gewicht : 0.99000kg
Artikelnummer. 10253570
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Beschreibung
Beschreibung
Glutaredoxin 2 (Hu) Recombinant Protein, LF-P0016, from Invitrogen.
Glutaredoxin (Grx), also known as thiol transferase, is a small heat-stable oxidoreductase. Grxs form part of the glutaredoxin system, comprising NADPH, GSH and glutathione reductase, which transfers electrons from NADPH to glutaredoxins via GSH. First discovered in E. coli as GSH-dependent hydrogen donors for ribonucleotide reductase, Grx catalyzes GSH-disulfide oxidoreductase via two redox-active cysteine residues. The active sequence (Cys-Pro-Tyr-Cys) is conserved in a variety of species. The 12-kD dithiol protein has a role in reduction of mixed disulfides in cells exposed to oxidative stress. This protein corresponds to human Glutaredoxin 2 (20-164 aa), which has a molecular weight of 14.1 kDa. Specific activity: 45 U/mg, where one unit will cause the oxidation of 1 umol of NADPH per min. Purity: >95% by SDS-PAGE.
Spezifikation
Spezifikation
Q9NS18
Control
51022
7.0
E. coli
RUO
GLRX2
Unconjugated
E. coli
Liquid
purified
1mg/mL
0.02M HEPES with 10% glycerol and no preservative; pH 7.0
Human GLRX2
0.1mg
-80°C, Avoid Freeze/Thaw Cycles
Human GLRX2
Human
Hu
Recombinant
Recombinant
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote
in vitro research only.