missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-16 Recombinant Protein

Recombinant Protein

Marke:  Invitrogen™ RP8610

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11529131

  • 337.00 € / 10 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.


Beschreibung

Beschreibung

IL-16 (mouse) Recombinant Protein, RP-8610, from Invitrogen.

Interleukin 16 (IL-16) is produced primarily by CD4+ and CD8+ T cells and acts as a chemo-attractant for lymphocytes, monocytes, eosinophils, dendritic cells and Langerhans cells.  Additionally, IL-16 has been reported to upregulate IL-2 receptor (CD25), induce progression of cells to the G1 phase and suppress HIV & SIV replication.   Recombinant mouse IL-16 is a non-glycosylated protein, containing 127 amino acids, with a molecular weight of 13.2 kDa. Recombinant mouse IL-16 has full biological activity compared to a standard. The activity, determined by dose-dependent chemoattraction of primary human T cells, is detectable starting at 5 ng/mL. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: MHDLNSSTDS AASASAASDI SVESKEATVC TVTLEKTSAG LGFSLEGGKG SLHGDKPLTI NRIFKGDRTG EMVQPGDEIL QLAGTAVQGL TRFEAWNVIK ALPDGPVTIV IRRTSLQCKQ TTASADS
Spezifikation

Spezifikation

O54824
Control
16170
Mouse IL-16
10μg
-20°C, Avoid Freeze/Thaw Cycles
M-IL-16
Il16
Unconjugated
E. coli
Lyophilized
purified
0.1mg/mL
10mM sodium phosphate with no preservative; pH 7.5
∽13.5
7.5
E. coli
RUO
Mouse IL-16
Mouse
Ms
Recombinant
Recombinant
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

in vitro research only.