Learn More
Invitrogen™ Mouse IL-7 Recombinant Protein
Recombinant Protein
Marke:  Invitrogen™ RP8664
Weitere Details : Netto-Gewicht : 0.01000kg
Artikelnummer. 11544970
  • 335.00 € / 10 Mikrogramm
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.
Beschreibung
Beschreibung
IL-7 (mouse) Recombinant Protein, RP-8664, from Invitrogen.
Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells. Recombinant mouse soluble RANK Ligand is a non-glycosylated protein, containing 174 amino acids, with a molecular weight of 19.9 kDa. Purity is typically greater than 98% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the ability to induce osteoclast formation on mouse RAW264.7 cells using a concentration of 5.0-10.0ng/mL. The specific activity of this protein is 2x10^5 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: PAMMEGSWLD VAQRGKPEAQ PFAHLTINAA SIPSGSHKVT LSSWYHDRGW AKISNMTLSN GKLRVNQDGF YYLYANICFR HHETSGSVPT DYLQLMVYVV KTSIKIPSSH NLMKGGSTKN WSGNSEFHFY SINVGGFFKL RAGEEISIQV SNPSLLDPDQ DATYFGAFKV QDID.
Spezifikation
Spezifikation
P10168
Control
16196
Mouse IL-7
E. coli
RUO
Mouse IL-7
Mouse
Ms
Recombinant
Recombinant
0.1mg/mL
0.1%TFA with no preservative
∽15
10μg
-20°C, Avoid Freeze/Thaw Cycles
M-IL-7
Il7
Unconjugated
E. coli
Lyophilized
purified
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote
in vitro research only