missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse CXCL10 (IP-10) Recombinant Protein
Recombinant Protein
Marke: Invitrogen™ RP8668
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
IP10 (mouse) Recombinant Protein, RP-8668, from Invitrogen.
The chemokine IP-10 (or CXCL10) is a chemokine made by monocytes, endothelial cells and fibroblasts in response to treatment with IFNgamma. IP-10 functions as a chemoattractant to cells expressing the G protein-coupled receptor, CXCR3, which is found mainly on activated T cells and NK cells. IP-10 plays an important role in Th1 type inflammatory diseases and autoimmune diseases such as, Hashimoto's thyroiditis, Graves' disease and Type 1 diabetes mellitus. Recombinant mouse MCP-1 is a non-glycosylated protein, containing 125 amino acids, with a molecular weight of 13.8 kDa. Purity is typically greater than 97% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the ability to chemoattract human PBMCs or THP-1 cells at 4-15 ng/mL. The specific activity of this protein is 2.5x10^5 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: QPDAVNAPLT CCYSFTSKMI PMSRLESYKR ITSSRCPKEA VVFVTKLKRE VCADPKKEWV QTYIKNLDRN QMRSEPTTLF KTASALRSSA PLNVKLTRKS EANASTTFST TTSSTSVGVT SVTVN.Spezifikation
P17515 | |
Control | |
15945 | |
Mouse CXCL10 | |
E. coli | |
RUO | |
Mouse CXCL10 | |
Mouse | |
Ms | |
Recombinant | |
Recombinant |
0.1mg/mL | |
0.1%TFA with no preservative | |
∽8.7 | |
25μg | |
-20°C, Avoid Freeze/Thaw Cycles | |
M-IP-10 | |
Cxcl10 | |
Unconjugated | |
E. coli | |
Lyophilized | |
purified |
in vitro research only