Learn More
Invitrogen™ Human CCL3 (MIP-1 alpha) Recombinant Protein
Recombinant Protein
Marke: Invitrogen™ RP10923
Weitere Details : CAS : 69-74-9 Netto-Gewicht : 0.01000kg
Beschreibung
The RP-10923 protein is a recombinant protein suitable for use as a control in ELISA and Western blot applications. Biological activity of RP-10923 determined by the ability to chemoattract human monocytes is 1-10ng/mL. Reconstitute the lyophilized product in sterile water not less than 100 μg/mL.
The RP-10923 protein is a recombinant protein suitable for use as a control in ELISA and Western blot applications. RP-10923 is reported to have the additional biological activity: Biological activity as determined by chemoattraction of human monocytes is 1-10ng/mL. Reconstitute the lyophilized product in sterile water not less than 100ug/mL.Spezifikation
P10147 | |
protein with no preservative | |
Human CCL3 (MIP-1 alpha) | |
E. coli | |
-20°C, Avoid Freeze/Thaw Cycles | |
MIP-1 alpha | |
CCL3 | |
Unconjugated | |
E. coli | |
Lyophilized | |
purified |
Control | |
6348 | |
20μg | |
N/A | |
RUO | |
Human CCL3 (MIP-1 alpha) | |
Human | |
Hu | |
Recombinant | |
Recombinant |
in vitro in vitro research only.