Learn More
Invitrogen™ Human MMP-14 (Prodomain-catalytic Domain) Recombinant Protein
Recombinant Protein
Marke: Invitrogen™ RP77532
Weitere Details : CAS : 69-74-9 Netto-Gewicht : 0.01000kg
Beschreibung
MMP14 Recombinant Protein, RP-77532, from Invitrogen.
RP-77532 is a purified recombinant MMP-14 (prodomain-catalytic domain) protein, expressed as a periplasmic protein in E. coli. The truncated proenzyme consists of MT1-MMP amino acid residues Ser1-Gly265 followed by the His-tag sequence Leu-Val-Thr-(His)6.Spezifikation
P50281 | |
TBS with 5mM CaCl2 and no preservative; pH 7.5 | |
Human MMP14 | |
10μg | |
-80°C, Avoid Freeze/Thaw Cycles | |
MT1-MMP | |
MMP14 | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
Control | |
4323 | |
7.5 | |
E. coli | |
RUO | |
Human MMP14 | |
Human | |
Hu | |
Recombinant | |
Recombinant |
in vitro research only