missing translation for 'onlineSavingsMsg'
Learn More

Phalloidin Control, DyLight 554, Invitrogen™

Control

Marke:  Invitrogen 21834

Weitere Details : Netto-Gewicht : 0.10000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : II

Artikelnummer. 13218118

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

Phalloidin Control, DyLight 554 conjugate, 21834, from Invitrogen., 21834, from Invitrogen. Species Reactivity: Many; Applications: Immunocytochemistry, Immunofluorescence, Immunohistochemistry.

Phalloidin is a bicyclic peptide that belongs to a family of toxins isolated from the deadly Amanita phalloides “death cap” mushroom and is commonly used as a counterstain (similar to DAPI or Hoechst) in cell biology and histology imaging applications to selectively label F-actin in fixed cells, permeabilized cells, and cell-free experiments. Labeled phalloidin conjugates have similar affinity for both large and small filaments and bind in a stoichiometric ratio of about one phallotoxin per actin subunit in both muscle and non-muscle cells. Phalloidins reportedly do not bind to monomeric G-actin, unlike some antibodies against actin. The dynamics of the actin polymerization in cells are important for a variety of cellular processes from cell motility to cell shape, from muscular contraction to cytokinesis, and more.
Cross Reactivity: Many Species
Formulation: 300 units purified control antibody (300 units/ml) in methanol and no preservative
Synonyms: DY554-Phalloidin, F-actin toxin, Amanita phalloides
Format: 300 units of lyophilized DyLight 554-Phalloidin (300 units/ml) in methanol. Prepare stock solution by adding 1000 µL of pure methanol to the vial and gently mix. 21834 has been successfully used in immunofluorescence and immunohistochemistry. DyLight 554-Phalloidin has an excitation/emission of 551/572nm and molecular weight of 1313.5 g/mole. DyLight 554-Phalloidin Stock Solution can be prepared in methanol, DMF, or DMSO and unused Stock Solution should be promptly stored at -20°C to minimize evaporation. Working solutions (per 96-well plate) can be prepared by diluting 20 µL of the Stock Solution in 5.98mL of PBS and mixing well. Typical staining procedure adds 50µl of Working Solution (i.e. 1 to 5 unit/ml) to a each well. Incubate cells in the dark for 30 minutes at room temperature (optimal staining times varies from 10 minutes to 3 hours depending on cell type). Aspirate and wash cells three times in PBS after incubation. If desired, probe with specific antibodies or dyes before using DyLight 554-Phalloidin.
Spezifikation

Spezifikation

Phalloidin
300 units/mL
DY554-Phalloidin, F-actin toxin, Amanita phalloides
300 U
Isotype Control (Primary)
-20°C
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
DyLight 554
purified
RUO
Multi-species
Lyophilized
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote