Learn More
Sheep IgG Isotype Control, Invitrogen™
Sheep Isotype Control
Marke: Invitrogen 31243
Weitere Details : Netto-Gewicht : 0.80000kg
Beschreibung
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Purity/Antibody specificity: Based on immunoelectrophoresis at an antigen concentration of 20 mg/mL, the pattern of precipitation against rabbit anti-sheep whole serum is the same as that against rabbit anti-sheep IgG, Fc fragment specific. This antibody is recommended for use in blocking assays, immunohistochemical, immunocytochemical and immunofluorescent applications. Store product at 4°C until opened. After opening, dilute only enough antibody for a single day's use. Store remainder at 4°C under sterile conditions. Country of Origin: USA
Sheep IgG Control antibody specifically detects Sheep IgG in Not Applicable samples. It is validated for Blocking Assay, Control, Immunocytochemistry, Immunohistochemistry.Spezifikation
Sheep IgG | |
11.9 mg/mL | |
PBS with no preservative; pH 7.6 | |
Sheep | |
10 mg | |
Isotype Control (Primary) | |
4° C | |
IgG |
Blocking Assay, Control, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry | |
Unconjugated | |
Immunoglobulin G; ImmunoglobulinG | |
purified | |
RUO | |
Not Applicable | |
Liquid |