missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Transthyretin, Recombinant Protein, Invitrogen™
Recombinant Protein
Marke: Invitrogen LFP0054
Weitere Details : Netto-Gewicht : 0.99000kg
Beschreibung
Transthyretin (Prealbumin) Recombinant Protein, LF-P0054, from Invitrogen.
Transthyretin is a constituent fTransthyretin (TTR) , generally called prealbumin, is a plasma protein that plays an important role in physiology such as a transporter of hormone thyroxine and retinal-binding protein. After produced primarily in the liver, TTR is excreted into the plasma. TTR represents a disproportionate fraction (25%) of CSF protein, prompting the suggestion that it is either selectively transported across the blood-CSF barrier or synthesized de novo within the central nervous system. Transthyretin is a constituent found to the neuritic plaques, neurofibrillary tangles, and microangiopathic lesions of senile cerebral amyloid. It has been reported that more than 40 different mutations in the TTR gene associated with amyloid deposition. This protein corresponds to human Prealbumin (21-147 aa), which has a molecular weight of 40.5 kDa. Purity: >95% by SDS-PAGE. After reconstitution, store at -80°C and avoid freeze and thaw cycles.Spezifikation
P02766 | |
Control | |
7276 | |
Human Transthyretin | |
500μg | |
-20°C | |
Human Transthyretin | |
Human | |
Hu | |
Recombinant | |
Recombinant |
1mg/mL | |
0.02M HEPES with no preservative; pH 7.0 | |
40.5 | |
7.0 | |
E. coli | |
RUO | |
TTR | |
Unconjugated | |
E. coli | |
Lyophilized | |
purified |
in vitro research only.