missing translation for 'onlineSavingsMsg'
Learn More

Thrombospondin-1 (TSP-1) Ab-2, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Artikelnummer. 12691377 Alle ansehen Epredia Produkte
Change view
Click to view available options
Menge:
100 μL
500 μL
1 mL
Konjugat:
Biotin
Unconjugated
Konzentration:
1mg/mL
200μg/mL
Unit Size:
1 Milliliter
100 Mikroliter
500 Mikroliter
Product Code. Menge Konjugat Konzentration unitSize
12691377 100 μL Biotin 200μg/mL 100 Mikroliter
12611387 500 μL Biotin 200μg/mL -
12601387 100 μL Unconjugated 200μg/mL 100 Mikroliter
12631387 500 μL Unconjugated 200μg/mL 500 Mikroliter
12621387 1 mL Unconjugated 200μg/mL 1 Milliliter
12641387 100 μL Unconjugated 1mg/mL 100 Mikroliter
6 options
This item is not returnable. View return policy

Product Code. 12691377

Brand: Epredia MS419B0

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
View alternative products

This item is not returnable. View return policy

Provide accurate, reproducible results with the Epredia™ Thrombospondin-1 (TSP-1) Ab-2, Mouse Monoclonal Antibody.

Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.

Host Species: Mouse

Clone: D4.6

Isotype: IgG1

Species Reactivity: Human, Mouse, Rat, Cow, Pig, Horse, Dog and Sheep. Others not known.

Epitope: Calcium-binding domain of TSP (C-terminal 50kDa piece of the 120kDa fragment)

Immunogen: Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets

Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)

Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunoprecipitation (Native verified)
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombospondin-1 (TSP-1) Ab-2
Anwendungen Immunoprecipitation, Western Blot
Klassifikation Monoclonal
Klon D4.6
Konzentration 200μg/mL
Konjugat Biotin
Wirtsspezies Mouse
Immunogen Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets
Menge 100 μL
Regulatorischer Status RUO
Forschungsgebiet Angiogenesis
Primär oder sekundär Primary
Zielspezies Bovine, Canine, Equine, Human, Mouse, Pig, Rat, Sheep
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.