Learn More
THUMPD1 Monoclonal Antibody (4A11), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA524483
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Peptide Sequence: MAAPAQQTTQ PGGGKRKGKA QYVLAKRARR CDAGGPRQLE PGLQGILITC NMNERKCVEE AYSLLNEYGD DMYGPEKFTD KDQQPSGSEG EDDDAEAALK KEVGDIKAST EMRLRRFQSV ESGANNVVFI RTLGIEPEKL VHHILQDMYK TKKKKTRVIL RMLPISGTCK AFLEDMKKYA ETFLEPWFKA PNKGTFQIVY KSRNNSHVNR EEVIRELAGI VCTLNSENKV DLTNPQYTVV VEIIKAVCCL SVVKDYMLFR KYNLQEVVKS PKDPSQLNSK QGNGKEAKLE SADKSDQNNT AEGKNNQQVP ENTEELGQTK PTSNPQVVNE GGAKPELASQ ATEGSKSNEN DFS
THUMPD1 Monoclonal antibody specifically detects THUMPD1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
THUMPD1 | |
Monoclonal | |
Unconjugated | |
Thumpd1 | |
6330575P11Rik; EGK_12570; Tan1; THUMP domain containing 1; THUMP domain-containing protein 1; THUMPD1 | |
Mouse | |
Protein A | |
RUO | |
55623 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4A11 | |
PBS with no preservative; pH 7.4 | |
Q9NXG2 | |
Thumpd1 | |
THUMPD1 (AAH00448, 1 a.a. ∽ 353 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |