missing translation for 'onlineSavingsMsg'
Learn More

Thymosin alpha 1 Antibody [DyLight 594], Novus Biologicals™

Product Code. 30753193 Shop All Bio Techne Products
Change view
Click to view available options
Menge:
0.1 mL
Unit Size:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30753193 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30753193 Supplier Novus Biologicals Supplier No. NBP346885DL594

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Thymosin alpha 1 Polyclonal antibody specifically detects Thymosin alpha 1 in Human samples. It is validated for ELISA,Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thymosin alpha 1
Anwendungen ELISA, Immunohistochemistry
Klassifikation Polyclonal
Konjugat DyLight 594
Zusammensetzung 50mM Sodium Borate
Gen-Alias alpha (gene sequence 28), gene sequence 28, MGC104802, prothymosin alpha, prothymosin alpha protein, prothymosin, alpha, PTMA prothymosin, alpha, TMSA
Wirtsspezies Rabbit
Immunogen prothymosin, alpha. (Uniprot# P06454)
Reinigungsverfahren Immunogen affinity purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Cell Cycle and Replication
Primär oder sekundär Primary
Gen-ID (Entrez) 5757
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.