missing translation for 'onlineSavingsMsg'
Learn More

Ticagrelor, TRC

Product Code. 30451857
Change view
Click to view available options
Menge:
50 mg
Unit Size:
50 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30451857 50 mg 50 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451857 Supplier Toronto Research Chemicals Supplier No. TRCT43770050MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Ticagrelor
CAS 274693-27-5
Summenformel C23 H28 F2 N6 O4 S
Menge 50 mg
Synonym (1S,2S,3R,5S)-3-[7-[[(1R,2S)-2-(3,4-difluorophenyl)cyclopropyl]amino]-5-(propylthio)-3H-1,2,3-triazolo[4,5-d]pyrimidin-3-yl]-5-(2-hydroxyethoxy)-1,2-cyclopentanediol, (1S,2S,3R,5S)-3-[7-[(1R,2S)-2-(3,4-Difluorophenyl)cyclopropyl]amino]-5-propylthio-3H-[1,2,3]triazolo[4,5-d]pyrimidin-3-yl]-5-(2-hydroxyethoxy)cyclopentane-1,2-diol, AR-C 126532XX, AZD 6140, Brilinta, Brilique, Ticagrelor
InChI-Formel InChI=1S/C23H28F2N6O4S/c1-2-7-36-23-27-21(26-15-9-12(15)11-3-4-13(24)14(25)8-11)18-22(28-23)31(30-29-18)16-10-17(35-6-5-32)20(34)19(16)33/h3-4,8,12,15-17,19-20,32-34H,2,5-7,9-10H2,1H3,(H,26,27,28)/t12-,15+,16+,17-,19-,20+/m0/s1
SMILES CCCSc1nc(N[C@@H]2C[C@H]2c3ccc(F)c(F)c3)c4nnn([C@@H]5C[C@H](OCCO)[C@@H](O)[C@H]5O)c4n1
IUPAC-Name (1S,2S,3R,5S)-3-[7-[[(1R,2S)-2-(3,4-Difluorphenyl)cyclopropyl]amino]-5-propylsulfanyltriazolo[4,5-d]pyrimidin-3-yl]-5-(2-hydroxyethoxy)cyclopentan-1,2-diol
Molekulargewicht (g/mol) 522.57
Formelmasse 522.1861
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.