missing translation for 'onlineSavingsMsg'
Learn More

TIM-3 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications

N.V.

Spezifikation

Antigen TIM-3
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TIM-3 Leu22-Arg191 Accession # AAL65156
Wirtsspezies Goat
Primär oder sekundär Primary
Klassifikation Polyclonal
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

TIM-3 Polyclonal specifically detects TIM-3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Spezifikation

Spezifikation

TIM-3
Goat
Polyclonal
IgG
CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Western Blot, Flow Cytometry, CyTOF
RUO
Mouse myeloma cell line NS0-derived recombinant mouse TIM-3 Leu22-Arg191 Accession # AAL65156
Primary
Mouse
Unconjugated
84868
Affinity Purified
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen

For Research Use Only