missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TIM-6 Antibody, R&D Systems™
Goat Polyclonal Antibody
N.V.
Specifications
Antigen | TIM-6 |
---|---|
Konzentration | LYOPH |
Verdünnung | Western Blot 0.1 ug/mL |
Anwendungen | Western Blot |
Klassifikation | Polyclonal |
Description
TIM-6 Polyclonal specifically detects TIM-6 in Mouse samples. It is validated for Western Blot.Specifications
TIM-6 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
Mouse myeloma cell line NS0-derived recombinant mouse TIM-6 Glu24-Ala140 Accession # NP_001020606 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse TIM-6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human TIM-3 and recombinant mouse TIM-4 is observed. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
NP_001020606 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only