Learn More
Invitrogen™ Human TIMP1, Recombinant Protein
Recombinant Protein
Marke:  Invitrogen™ RP75733
Weitere Details : CAS : 69-74-9 Netto-Gewicht : 0.01000kg
Artikelnummer. 11529051
  • 243.00 € / 10 Mikrogramm
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.
Beschreibung
Beschreibung
TIMP1 (human) Recombinant Protein, RP-75733, from Invitrogen.
TIMP1 (metalloproteinase inhibitor 1) is a metalloproteinase inhibitor that functions by forms one-to-one complexes with target metallproteinases, such as collagenases, and irreversible inactivates them by binding to their catalytic zinc cofactor. TIMP1 acts on MMP1, MMP2, MMP3, MMP7, MMP8, MMP9, MMP10, MMP11, MMP12, MMP14, and MMP16. It also functions as a growth factor that regulates cell differenation, migration, and cell death. TIMP1 activates cellular signaling cascades via CD63 and ITGB1. This product contains a purified 184 amino acid fragment (aa24-207) corresponding to the mature TIMP-1 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays. This protein has a molecular weight of ∽25.21 kDa.
Spezifikation
Spezifikation
P01033
Control, Western Blot
7076
4.8
E. Coli
RUO
Human TIMP1
Human
Hu
Recombinant
Recombinant
0.31mg/mL
0.025M acetic acid with 50% glycerol and no preservative; pH 4.8
Human TIMP1
10μg
-20°C, Avoid Freeze/Thaw Cycles
Tissue Inhibitor of Metalloproteinase 1,TIMP1
TIMP1