missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
Antibody detects endogenous levels of total TIRAP.
Toll-like receptors (TLRs) are signaling molecules that recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses. These proteins act through adaptor molecules such as TIRAP and MyD88 to activate various kinases and transcription factors. In TIRAP-deficient mice, TLR signaling in response to TLR2 ligands (using either TLR1 and TLR6 as co-receptors) is totally abolished, suggesting that MyD88 and TIRAP work together and are both required for TLR2 signaling. Furthermore, these mice are also resistant to the toxic effects of LPS and show defects in NF-βB and MAP kinase activation, suggesting that TIRAP is also involed in TLR4 signaling.
Spezifikation
Spezifikation
| Antigen | TIRAP |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.02% sodium azide |
| Gen | TIRAP |
| Gen-Zugriffsnummer | P58753 |
| Gen-Alias | AA407980; adapter protein wyatt; adaptor protein Wyatt; BACTS1; C130027E04Rik; FLJ42305; HGNC:17192; Mal; myD88 adapter-like protein; MyD88-2; OTTHUMP00000179130; TIR domain containing adaptor protein; TIR domain-containing adapter protein; TIRAP; Tlr4ap; toll/interleukin-1 receptor domain-containing adapter protein; toll-interleukin 1 receptor (TIR) domain containing adaptor protein; toll-interleukin 1 receptor (TIR) domain-containing adaptor protein; Toll-like receptor 4 adaptor protein; Toll-like receptor adaptor protein; Wyatt |
| Gensymbole | TIRAP |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?
