missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TIS11D Polyclonal specifically detects TIS11D in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | TIS11D |
| Anwendungen | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | DyLight 650 |
| Verdünnung | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | HF.10, HF10Zinc finger protein HF.10, Zfp105, zinc finger protein 35, zinc finger protein 35 (clone HF.10) |
| Wirtsspezies | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human TIS11D fragment. |
| Reinigungsverfahren | Antigen Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?