missing translation for 'onlineSavingsMsg'
Learn More

TLR7 Antibody (4G6) - Azide and BSA Free, Novus Biologicals™

Product Code. 18103943 Shop All Bio Techne Products
Change view
Click to view available options
Menge:
0.025 mg
Unit Size:
0.10 Milligramm
25 Mikrogramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
18103943 0.025 mg 25 Mikrogramm
18777743 - 0.10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18103943 Supplier Novus Biologicals Supplier No. NBP225274SS

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

TLR7 Monoclonal specifically detects TLR7 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount, CyTOF-ready, Knockdown Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR7
Anwendungen Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), CyTOF, KnockDown
Klassifikation Monoclonal
Klon 4G6
Konzentration 1.0 mg/mL
Konjugat Unconjugated
Verdünnung Western Blot 10 ug/ml, Flow Cytometry 1ul/1 million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500-1:5000. Use reported in scientific literature (PMID 22610069), Immunohistochemistry-Frozen reported in scientific literature (PMID 22610069), Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 22610069), CyTOF-ready, KnockDown Validated
Gen-Zugriffsnummer NP_057646
Gen-Alias toll-like receptor 7
Gensymbole TLR7
Wirtsspezies Mouse
Immunogen A partial human TLR7 recombinant protein (amino acids 562-839) was used as the immunogen.
Reinigungsverfahren Protein G purified
Menge 0.025 mg
Regulatorischer Status RUO
Forschungsgebiet Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
Primär oder sekundär Primary
Gen-ID (Entrez) 51284
Testspezifität This product reacts with Human.
Zielspezies Human, Mouse
Inhalt und Lagerung Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.