missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Beskrivning
Immunogen sequence: IPPYVMKCPS NGLCSRLPAD CIDCTTNFSC TYGKPVTFDC AVKPSVTCVD QDFKSQKNFI INMTCRFCWQ LPETDYECTN STSCMTVSC Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 79%.
The protein encoded by this gene contains a structural module related to that of the seven transmembrane domain G protein-coupled receptor superfamily. This protein has sequence and structural similarities to the beta-amyloid binding protein (BBP), but, unlike BBP, it does not regulate a response to beta-amyloid peptide. This protein may have regulatory roles in cell death or proliferation signal cascades. Several alternatively spliced transcript variants of this gene are described but the full length nature of some variants has not been determined. Multiple polyadenylation sites have been found in this gene.
Specifikationer
Specifikationer
| Antigen | TM2D3 |
| Anwendungen | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Klassifikation | Polyclonal |
| Konzentration | 0.10 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gen | TM2D3 |
| Gen-Zugriffsnummer | Q9BRN9 |
| Gen-Alias | 1110025I09Rik; 5930422O05Rik; BBP-like protein 2; beta-amyloid-binding protein-like protein 2; Blp2; fc17h12; LOW QUALITY PROTEIN: TM2 domain-containing protein 3; RGD1564929; TM2 domain containing 3; TM2 domain-containing protein 3; tm2d3; wu:fc17h12; ZFOS-508H5.2; zgc:165591 |
| Gensymbole | TM2D3 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?
