missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM70 Mouse anti-Human, Mouse, Clone: 2C11C3, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 60195-1-IG-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
TMEM70 is a recently identified mitochondrial protein that is thought to play a role in the biogenesis of the ATP synthase in higher eukaryotes. Mutations in this gene result in early neonatal onset of hypotonia, hypertrophic cardiomyopathy, lactic acidos and 3-methylglutaconic aciduria (3-MGC-uria), and usually cause death within the first six weeks of life, although some patients survive much longer. Little is known of the role of TMEM70, but it is conserved across multicellular eukaryotic organisms. It contains a conserved DUF1301 domain and two putative transmembrane regions.Spezifikation
TMEM70 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q921N7, Q9BUB7 | |
TMEM70 | |
TMEM70 Fusion Protein Ag16947 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
2C11C3 | |
Unconjugated | |
TMEM70 | |
TMEM70, transmembrane protein 70 | |
Mouse | |
Protein A | |
RUO | |
54968, 70397 | |
-20°C | |
Liquid |