missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMOD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10145-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
TMOD1 gene encodes a member of the tropomodulin family. The encoded protein is an actin-capping protein that regulates tropomyosin by binding to its N-terminus, inhibiting depolymerization and elongation of the pointed end of actin filaments and thereby influencing the structure of the erythrocyte membrane skeleton. Multiple transcript variants encoding the same protein have been found for this gene.Spezifikation
TMOD1 | |
Polyclonal | |
Unconjugated | |
TMOD1 | |
D9S57E, E Tmod, Erythrocyte tropomodulin, ETMOD, TMOD, TMOD1, tropomodulin 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21916, 25566, 7111 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P28289, P49813, P70567 | |
Tmod1 | |
TMOD1 Fusion Protein Ag0192 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |