missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMPRSS5 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 24616-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a protein that belongs to the serine protease family. Serine proteases are known to be involved in many physiological and pathological processes.Spezifikation
TMPRSS5 | |
Polyclonal | |
Unconjugated | |
TMPRSS5 | |
SPINESIN, TMPRSS5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
266681, 80975 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H3S3 | |
TMPRSS5 | |
TMPRSS5 Fusion Protein Ag18493 | |
20 μL | |
Primary | |
Rat, Human | |
Antibody | |
IgG |