missing translation for 'onlineSavingsMsg'
Learn More

TNFSF13 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 10680-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16887213

  • 329.85 € / 150 Mikroliter

Voraussichtliches Versanddatum: 23-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Members in the TNF superfamily regulate immune responses and induce apoptosis. A novel member in the TNF family was recently identified by several groups and designated APRIL (for A PRoliferation-Inducing Ligand), TALL2 (for TNF and ApoL-related Leukocyte-expressed Ligand 2), and TRDL 1a (for TNF related death ligand 1a) in human and mouse. Two receptors for APRIL were recently identified and designated TACI and BCMA. APRIL stimulates B and T cell proliferation, triggers humoral immune responses, activates NF-kB, and induces cell death. APRIL and its close relative of BlyS and their receptors BCMA and TACI are involved in diseases of autoimmunity and cancer.
Spezifikation

Spezifikation

TNFSF13
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O75888
APRIL, CD256, ligand, TALL 2, TALL2, TNF related death ligand 1, TNFSF13, TRDL 1, UNQ383/PRO715, ZTNF2
Rabbit
IgG
150 μL
-20°C
Primary
8741
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TNFSF13
Liquid
TNFSF13
TNFSF13 Fusion Protein Ag1056
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate