missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toltrazuril-d3, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Toltrazuril D3 (N-methyl D3) |
| CAS | 1353867-75-0 |
| Summenformel | C18 2H3 H11 F3 N3 O4 S |
| Menge | 1 mg |
| Synonym | 1-[3-Methyl-4-[4-(trifluoromethylsulfanyl)phenoxy]phenyl]-3-(trideuteriomethyl)-1,3,5-triazinane-2,4,6-trione |
| InChI-Formel | InChI=1S/C18H14F3N3O4S/c1-10-9-11(24-16(26)22-15(25)23(2)17(24)27)3-8-14(10)28-12-4-6-13(7-5-12)29-18(19,20)21/h3-9H,1-2H3,(H,22,25,26)/i2D3 |
| SMILES | [2H]C([2H])([2H])N1C(=O)NC(=O)N(C1=O)c2ccc(Oc3ccc(SC(F)(F)F)cc3)c(C)c2 |
| IUPAC-Name | 1-[3-Methyl-4-[4-(Trifluormethylsulfanyl)phenoxy]phenyl]-3-(trideuteriomethyl)-1,3,5-triazinan-2,4,6-trion |
| Molekulargewicht (g/mol) | 428.4 |
| Formelmasse | 428.085 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?