Learn More
Toluene D8 99.50% D, Eurisotop
Product Code. 17247366
Encompass
Change view
Click to view available options
Quantity:
10 mL
2 x 0.75 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
17247366 10 mL
17227366 2 x 0.75 mL
17237366 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17247366 Supplier Eurisotop Supplier No. D005FE
 Restricted Item
Verfügbar ab: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Synonym: Toluene-d8
Toluene D8
  • Isotopic Enrichment: 99.50% D
CoA and MSDS available at Eurisotop.com
Chemical Identifiers
CAS 2037-26-5
Molecular Formula C7H8
Molecular Weight (g/mol) 100.19
InChI Key YXFVVABEGXRONW-JGUCLWPXSA-N
Synonym Toluene-d8
IUPAC Name 1-(2H3)methyl(2H5)benzene
SMILES [2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])C([2H])([2H])[2H]
Specifications
Color Colorless
Deuterium (D) 99.50% D
Quantity 10 mL
Melting Point -95°C
Boiling Point 110.6°C
Grade Research
Chemical Name or Material Toluene D8