Learn More
Toluene D8 99.94% D, Eurisotop
Description
Toluene D8
- Isotopic Enrichment: 99.94% D
- Quantity: (2) 0.75 mL
- Packaging: Ampule
- Water Content: water <0.02%
CoA and MSDS available at Eurisotop.com
Specifications
Specifications
| Color | Colorless |
| Deuterium (D) | 99.94% D |
| Packaging | Ampule |
| Quantity | 2 x 0.75 mL |
| CAS | 2037-26-5 |
| Melting Point | -95°C |
| Boiling Point | 110.6°C |
| Molecular Formula | C7H8 |
| Synonym | Toluene-d8 |
| InChI Key | YXFVVABEGXRONW-JGUCLWPXSA-N |
| Show More |