Learn More
Tolyltriazole, TRC
Product Code. 30453439
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453439 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30453439 Supplier Toronto Research Chemicals Supplier No. TRCT5370001G
 Restricted Item
Verfügbar ab: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tolyltriazole
CAS 29385-43-1
Purity Grade Notes HPLC
Molecular Formula 2 C7 H7 N3
Quantity 1 g
Synonym 1H-Benzotriazole','6(or 7)-methyl-, 1H-Benzotriazole','4(or 5)-methyl- (9CI), 6(or 7)-Methyl-1H-benzotriazole, Additin RC 8221, C.V.I. TT, Cobratec TT 100, Cobratec TT 35I, Iramet 39, Methylbenzotriazole, Preventol CI 7-100, Seetec T, Seetec TT, Seetec TT-R, Stabinol MBTZ, TTZ, Tolyltriazole
InChI Formula InChI=1S/2C7H7N3/c1-5-2-3-6-7(4-5)9-10-8-6;1-5-3-2-4-6-7(5)9-10-8-6/h2*2-4H,1H3,(H,8,9,10)
SMILES Cc1ccc2[nH]nnc2c1.Cc3cccc4[nH]nnc34
IUPAC Name 4-methyl-1H-benzotriazole;5-methyl-1H-benzotriazole
Molecular Weight (g/mol) 266.3
Formula Weight 266.128
Percent Purity >95