missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOX Monoclonal Antibody (TXRX10), eBioscience™

Artikelnummer. 17198525
Change view
Click to view available options
Menge:
10 mg
100 μg
2 mg
25 μg
Packungsgröße:
10 Milligramm
100 Mikrogramm
2 Milligramm
25 Mikrogramm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17198525 2 mg 2 Milligramm
15237717 25 μg 25 Mikrogramm
15247717 100 μg 100 Mikrogramm
17108535 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17198525 Lieferant Invitrogen™ Lieferanten-Nr. 14650237

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rat Monoclonal Antibody

Description: This TXRX10 monoclonal antibody reacts with human and mouse TOX, which is also known as Thymocyte selection-associated HMG box protein. This DNA-binding nuclear factor is expressed in thymocytes, T lymphocytes, NK cells, and lymphoid tissue-inducer (LTi) cells. TOX expression in thymocytes is upregulated during beta-selection and positive selection. Studies using Tox knockout mice have demonstrated a critical role for TOX in CD4^+ T cell development as a regulator of the CD4^loCD8^lo to CD4^+CD8^lo transition. Moreover, TOX has been shown to be involved in the development of NK and LTi cells. While its regulation remains unclear, TOX expression has been shown to be induced by calcineurin-mediated TCR signaling in thymocytes during positive selection. Applications Reported: This TXRX10 antibody has been reported for use in immunoprecipitation and immunoblotting (WB). Applications Tested: This TXRX10 antibody has been tested by immunoblotting of reduced cell lysate prepared from mouse thymocytes at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

TOX (thymocyte selection-associated high mobility group (HMG) box protein) is a member of the HMG box family of DNA-binding proteins and likely plays a role in the regulation of T-cell development. It is a 526 amino acid nuclear protein and the expression of TOX is upregulated by pre-T cell receptor (pre-TCR) and TCR activation in immature thymocytes. TOX-mediated positive selection is associated with up-regulation of RUNX3 and is calcineurin dependent. TOX-dependent transition to the CD4+CD8 stage is required for development of class II major histocompatibility complex-specific T cells.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen TOX
Anwendungen Immunoprecipitation, Western Blot
Klassifikation Monoclonal
Klon TXRX10
Konzentration 0.5 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 0.09% sodium azide; pH 7.2
Gen TOX
Gen-Zugriffsnummer O94900, Q66JW3
Gen-Alias 1700007F02Rik; KIAA0808; thymocyte selection associated high mobility group box; thymocyte selection-associated high mobility group box; thymocyte selection-associated high mobility group box protein TOX; thymocyte selection-associated HMG box; thymus high mobility group box protein TOX; TOX; TOX1
Gensymbole TOX
Wirtsspezies Rat
Reinigungsverfahren Affinity chromatography
Menge 2 mg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 252838, 9760
Zielspezies Human, Mouse
Inhalt und Lagerung 4°C
Produkttyp Antibody
Form Liquid
Isotype IgG2a κ
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.