missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
TRAIP Polyclonal antibody specifically detects TRAIP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Spezifikation
Spezifikation
| Antigen | TRAIP |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | PerCP |
| Verdünnung | Immunohistochemistry-Paraffin |
| Zusammensetzung | PBS |
| Gen-Alias | RNF206TRIPRING finger protein 206, TRAF interacting protein, TRAF-interacting protein |
| Wirtsspezies | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human TRAIP fragment. |
| Reinigungsverfahren | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?