missing translation for 'onlineSavingsMsg'
Learn More

Trans-3-Aminocyclohexanecarboxamid-Hydrochlorid, 97 %, Thermo Scientific Chemicals

Catalog Number 15437536
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
920966-29-6
C7H15ClN2O
178.66
MFCD12022628
ZCFBGXVDLNNMDE-KGZKBUQUSA-N
trans-3-aminocyclohexanecarboxamide hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxamide hydrochloride
46172883
(1R,3R)-3-Aminocyclohexan-1-carboxamid;hydrochlorid
C1CC(CC(C1)N)C(=O)N.Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
920966-29-6
C7H15ClN2O
178.66
MFCD12022628
ZCFBGXVDLNNMDE-KGZKBUQUSA-N
trans-3-aminocyclohexanecarboxamide hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxamide hydrochloride
46172883
(1R,3R)-3-Aminocyclohexan-1-carboxamid;hydrochlorid
C1CC(CC(C1)N)C(=O)N.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material trans-3-Aminocyclohexanecarboxamide hydrochloride
Formelmasse 178.65
Reinheit (%) 97%
Menge 1 g

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'