missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-(6,8-Dibrom-1,4-dihydrochinazolin-3(2H)-yl)cyclohexanolhydrochlorid, LoGiCal
Spezifikation
Spezifikation
| Chemischer Name oder Material | trans-4-(6,8-Dibromo-1,4-dihydroquinazolin-3(2H)-yl)cyclohexanol Hydrochloride |
| CAS | 15942-08-2 |
| Bezeichnung der Analyten oder Bestandteile | trans-4-(6,8-Dibromo-1,4-dihydroquinazolin-3(2H)-yl)cyclohexanol Hydrochloride |
| Versandbedingung | Room Temperature |
| Summenformel | C14 H18 Br2 N2 O . Cl H |
| Zertifizierung/Konformität | ISO 9001 |
| Synonym | Ambroxol Hydrochloride Imp. B (EP), Ambroxol Imp. B (EP), trans-4-(6,8-Dibromo-1,4-dihydroquinazolin-3(2H)-yl)cyclohexanol Hydrochloride, Ambroxol Hydrochloride Imp. B (EP) as Hydrochloride, trans-4-(6,8-Dibromo-1,4-dihydro-3(2H)-quinazolinyl)cyclohexanol monohydrochloride, Ambroxol Hydrochloride Impurity B as Hydrochloride, Ambroxol Impurity B as Hydrochloride, trans-4-(6,8-Dibromo-1,4-dihydro-3(2H)-quinazolinyl)cyclohexanol Hydrochloride, NA 873 Hydrochloride, Ambroxol EP Impurity B Hydrochloride, Ambroxol Cyclic Impurity Hydrochloride |
| InChI-Formel | InChI=1S/C14H18Br2N2O.ClH/c15-10-5-9-7-18(8-17-14(9)13(16)6-10)11-1-3-12(19)4-2-11;/h5-6,11-12,17,19H,1-4,7-8H2;1H/t11-,12-; |
| Lösungstyp | Neat |
| SMILES | Cl.O[C@@H]1CC[C@H](CC1)N2CNc3c(Br)cc(Br)cc3C2 |
| Mehr anzeigen |
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?